Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDP05602506 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 kg
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDP05602506 250 g Natural Poly Bottle
RDCP0560100 100 g White Poly Wide Mouth Bottle
RDCP0560500 500 g White Poly Wide Mouth Bottle
RDCP05603A5 3 kg Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RDP05602506 Supplier Ricca Chemical Company Supplier No. RDCP0560250A6
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point ∼1330°C
Melting Point ∼0°C
Color White
CAS Min % 100
pH 6.00 to 9.20
CAS Max % 100
Physical Form Solid
Quantity 250 g
Formula Weight 166
Percent Purity ≥ 99%
Grade Laboratory
Packaging Natural Poly Bottle
Density 3.12
Chemical Name or Material Potassium Iodide, ACS Reagent Grade
Show More Show Less
ShelfLife 84 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.