Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granular, FCC, 99-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18605619 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-619 2.5 kg Amber Glass Bottle
18-605-616 125 g Amber Glass Bottle
18-605-617 500 g Amber Glass Bottle
18-605-618 6 x 500 g Amber Glass Bottle
18-605-620 4 x 2.5 kg Amber Glass Bottle
18-605-621 12 kg Poly Pail
18-605-622 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-619 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P13442.5KG
Add to Cart
Add to Cart

P1344, 7681-11-0, KI

Spectrum™ Chemical Potassium Iodide, Granular, FCC is used as a source for dietary iodine. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 101.5%
Grade FCC
Loss on Drying 0.01
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.