Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice

Catalog No. 6002258 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
6 x 100 g
6 x 250 g
6 x 500 g
6 x 1 kg
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-58 250 g
60-022-56 100 g
60-045-812 6 x 100 g
60-045-814 6 x 250 g
60-045-815 6 x 500 g
60-022-60 5 kg
60-022-59 500 g
60-022-57 1 kg
60-045-813 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-022-58 Supplier Solstice Supplier No. 30315250G
Add to Cart
Add to Cart

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 250 g
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Vapor Pressure 1mmHg (745°C)
Chemical Name or Material Potassium Iodide, Reag. Ph. Eur. reag. ISO
missing translation for 'healthHazard2' P305 + P351 + P338
missing translation for 'healthHazard3' H302-H315-H319
EINECSNumber 231-659-4
RTECSNumber TT2975000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.