Learn More
Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 1,323°C |
| Melting Point | 681°C (literature) |
| CAS Max % | 100.0000% |
| Quantity | 6 x 100 g |
| Linear Formula | KI |
| Formula Weight | 166 |
| Percent Purity | >99.5% |
| Grade | puriss. p.a. |
| Packaging | Poly Bottle |
| Density | 3.12 g/cm3 |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| EINECSNumber | 231-659-4 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.