Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium (iodomethyl)trifluoroborate, tech. 90%

Catalog No. AAH6383903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6383903 1 g
AAH6383906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6383903 Supplier Thermo Scientific Chemicals Supplier No. H6383903

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

CAS: 888711-47-5 | CH2BF3IK | 247.83 g/mol

Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.

Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 888711-47-5
Molecular Formula CH2BF3IK
Molecular Weight (g/mol) 247.83
MDL Number MFCD11112187
InChI Key WQJVWRYIXQXISQ-UHFFFAOYSA-N
Synonym potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
PubChem CID 23674071
SMILES [K+].F[B-](F)(F)CI

Specifications

Melting Point 242°C to 258°C
Color Brown to White
Physical Form Solid
Quantity 1 g
Linear Formula ICH2BF3K
IUPAC Name potassium;trifluoro(iodomethyl)boranuide
Formula Weight 247.83
Percent Purity 90%
Grade Technical
Sensitivity Light sensitive
Chemical Name or Material Potassium (iodomethyl)trifluoroborate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.