Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, 10% (w/v), Ricca Chemical
SDP

Catalog No. 63174 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6317-4 120 mL Natural Poly Bottle
6317-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6317-4 Supplier Ricca Chemical Company Supplier No. 63174
 Restricted Item
Add to Cart
Add to Cart

Filling Solution for Cupric, Lead, Silver/Sulfide, and Thiocyanate Ion Selective Electrodes (ISE)

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Percent Purity 10%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
pH ∼7
Quantity 120 mL
Chemical Name or Material Potassium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.