Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, 99.0% min., ACS, MilliporeSigma™

Catalog No. MPX15205 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX15205 2.5 kg
MPX15201 500 g
MPX15207 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MPX15205 Supplier MilliporeSigma Supplier No. PX15205
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 99.0% min.
Linear Formula KNO3
Formula Weight 11.1
Grade EP/ISO/Reagent
Density 2.11g/cm3 (20°C); Bulk Density: 800 kg/m3
Chemical Name or Material Potassium Nitrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.