Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium nitrate, 99.994% (metals basis)

Catalog No. p-7048606
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1100814 25 g
AA1100822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1100814 Supplier Thermo Scientific Chemicals Supplier No. 01100814
Only null left
Add to Cart

CAS: 7757-79-1 | KNO3 | 101.10 g/mol

Potassium nitrate has several applications. It is used as an oxidizer, food preservative, fertilizer, and an active ingredient of condensed aerosol fire suppression systems. It is a powerful nitrating agent in sulfuric acid medium. In addition, it also used as an electrolyte in a salt bridge and thermal storage medium in power generation systems. Ternary salts containing potassium nitrate have been reported to improve the heat storage capacity in the molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400°C (decomposition)
Melting Point 334°C
Color White
Physical Form Crystalline
Quantity 25 g
UN Number UN1486
Merck Index 14,7648
Solubility Information Freely soluble in water. Slightly soluble in ethanol.
IUPAC Name potassium;nitrate
Formula Weight 101.11
Percent Purity 99.994%
Odor Odorless
Sensitivity Hygroscopic
Density 2.109 g/mL
Assay (metals basis)
Chemical Name or Material Potassium nitrate
Show More Show Less
Hazard Category H272
Hazard Statement GHS H Statement
H272
May intensify fire
oxidizer.
Precautionary Statement P210-P220-P221-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM NITRATE
EINECSNumber 231-818-8
RTECSNumber TT3700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.