Potassium nitrate, 99.997% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer: Alfa Aesar 1084430

Quantity 250g

 View more versions of this product

Catalog No. AA1084430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7681-11-0
    Physical Form Powder
    Assay Percent Range 99.998% (metals basis)
    Molecular Formula KI
    Formula Weight 166.01
    MDL Number MFCD00011405
    Chemical Name or Material Potassium nitrate

    Specifications

    Formula Weight 166.01
    Quantity 250g
    Color White
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.998% (metals basis)
    Molecular Formula KI
    Appearance White crystalline powder
    MDL Number MFCD00011405
    CAS 7681-11-0
    Vapor Pressure 1.33 hpa (1mm Hg) at 745°C
    Chemical Name or Material Potassium nitrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit