Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium nitrate, ACS, 99.0% min

Catalog No. p-4407752
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1344322 100 g
AA1344336 500 g
AA13443A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1344322 Supplier Thermo Scientific Chemicals Supplier No. 01344322
Only null left
Add to Cart

For Pyrotechnics, Glass Manufacture, Tempering Steel | CAS: 7757-79-1 | KNO3 | 101.10 g/mol

Pyrotechnics, glass manufacture, tempering steel, reagentPotassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.0% min.
IUPAC Name potassium nitrate
Formula Weight 101.11
Grade ACS
Odor Odorless
Appearance White crystalline
Density 2.109g/cm3
Chemical Name or Material Potassium nitrate, For ACS analysis
Show More Show Less
Hazard Category H272-H303: May intensify fire; oxidizer. May be harmful if swallowed.
DOTInformation Hazard Class: 5.1, Packing Group: III
EINECSNumber 231-818-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.