Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18612803 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-803 50 kg Poly Bottle
18-612-798 25 g Poly Bottle
18-612-799 125 g Poly Bottle
18-612-800 500 g Poly Bottle
18-612-801 2.5 kg Poly Bottle
18-612-802 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-612-803 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P134550KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

pH 4.5 to 8.5
CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Percent Purity 99%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Nitrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.