Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, Crystal, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605659 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-659 2.5 kg Poly Pail
18-605-660 12 kg Poly Pail
18-605-661 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-605-659 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P18432.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1843, 7757-79-1, KNO3

Spectrum™ Chemical Potassium Nitrate, Crystal, USP is used as a diuretic and diaphoretic. All Spectrum Chemical USP products, including all USP potassium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 100.5%
Grade USP
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.