Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, Purified, Reagents

Catalog No. 01337096
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-096 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-096 Supplier Reagents Holdings Llc Supplier No. C2286500500A5
Add to Cart
Add to Cart

Potassium Nitrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym Saltpeter, Nitre, Nitrate of Potash
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Boiling Point 400°C
Melting Point 334°C
Color White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.11
Chemical Name or Material Potassium Nitrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.