Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, TraceSELECT™, ≥99.995% (metals basis), Solstice

Catalog No. 60047180 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-180 25 g Poly bottle
60-047-179 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-180 Supplier Solstice Supplier No. 6042925G

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Melting Point 335°C (1013hPa)
Color Colorless
pH 5.5 to 7.5
Physical Form Crystalline
Quantity 25 g
UN Number UN1486
Formula Weight 101.1g/mol
Percent Purity ≥99.995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly bottle
Density 2.11g/cm3
Chemical Name or Material Potassium nitrate
Show More Show Less
Hazard Statement P220
DOTInformation 5.1 / PGIII
RTECSNumber 231-818-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.