Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrite, ≥96.0%, ACS Reagent, Solstice

Catalog No. 60045885 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-885 500 g Poly Bottle
60-045-884 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-045-885 Supplier Solstice Supplier No. 310484500G
This item is not returnable. View return policy

ACS Reagent,≥96.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
IUPAC Name potassium;nitrite
SMILES [K+].[O-]N=O

Specifications

Melting Point 350°C
Quantity 500 g
UN Number UN1488
Formula Weight 85.1g/mol
Percent Purity ≥96.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.920g/cm3
Chemical Name or Material Potassium Nitrite
Hazard Statement P220-P273-P301 + P310
Precautionary Statement H272-H301-H400
DOTInformation 5.1/PGII
EINECSNumber 231-832-4
RTECSNumber TT3750000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.