Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrite (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. P267250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P267-250 250 g Glass Bottle
P267-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. P267-250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Melting Point 387°C
Color White
pH 7 to 10
Physical Form Solid
Quantity 250 g
Assay Percent Range ≥96 %
UN Number 1488
Formula Weight 85.1
Percent Purity ≥96%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 1.9150g/cm³
Chemical Name or Material Potassium Nitrite
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.