Learn More
Selleck Chemical LLC Potassium oxonate-E0353-500MG
SDP
Supplier:  Selleck Chemical LLC E0353500MG
Encompass_Preferred
Potassium oxonate (Oxonic Acid Potassium Salt) is a selectively competitive uricase inhibitor produced hyperuricemia (HUA) in rodents
Catalog No. 50-313-6833
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.