Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.100 N (N/10), 0.0200 M (M/50), Ricca Tightrant™
SDP

Catalog No. 640032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Safety-coated Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6400-32 1 L Amber Glass Bottle
640016 500 mL Amber Glass Bottle
6400-1 4 L Amber Glass Bottle
R64000004F 4 L Cubitainer
6400-1SC 4 L Safety-coated Glass Bottle
6400-2-5 10 L Cubitainer
6400-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 6400-32 Supplier Ricca Chemical Company Supplier No. 640032
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

CAS Min % 98.0, 0.31
CAS Max % 100.0, 0.32
Grade ACS/ASTM E 200/AOAC/EPA
Concentration or Composition (by Analyte or Components) 0.100N, 0.0200M
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Potassium Permanganate, 0.100N, USP Volumetric Solution
ShelfLife The 10L (640025) and 20L (64005) sizes have six-month expiration (others, 18 months) due to package type.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.