Learn More
Potassium permanganate, 0.1N Standardized Solution
Description
Potassium permanganate, 0.1N standardized solution is used for volumetric analysis in laboratories. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Potassium permanganate |
| Color | Purple |
| CAS | 7722-64-7 |
| Physical Form | Liquid |
| Quantity | 4 L |
| Molecular Formula | KMnO4 |
| MDL Number | MFCD00011364 |
| Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
| Solubility Information | Soluble in water. |
| InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.