Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 5% (w/v), Mercury Free, Ricca Chemical
SDP

Catalog No. 636025 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
63602-5 10 L Cubitainer
6360-16 500 mL Amber Glass Bottle
6360-32 1 L Amber Glass Bottle
6360-1 4 L Amber Glass Bottle
R636000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 63602-5 Supplier Ricca Chemical Company Supplier No. 63602.5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Packaging Cubitainer
Concentration or Composition Notes 5% (w/v)
CAS Min % 94.0, 3.89
CAS Max % 98.0, 4.05
Physical Form Liquid
Color Purple
Melting Point Approximately 0°C
Solvent or Matrix Water
Quantity 10 L
Chemical Name or Material Potassium Permanganate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.