Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 891mg/L Equivalent to 1000 ppm Chlorine, Reagents

Catalog No. 01334177
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-177 500 mL
01-334-178 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-177 Supplier Reagents Holdings Llc Supplier No. CP208100500C
Add to Cart
Add to Cart

Potassium Permanganate, 891mg/L Equivalent to 1000ppm Chlorine, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7722-64-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Glass Bottle
Specific Gravity 1.0006
Physical Form Liquid
Color Purple
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 500 mL
Percent Purity 0.09%
Grade Reagent
Chemical Name or Material Potassium Permanganate, 891 mg/L Equivalent to 1000 ppm Chlorine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.