Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, ACS Reagent, Solstice

Catalog No. 6001297 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-012-97 1 kg
60-012-98 25 g
60-012-99 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-012-97 Supplier Solstice Supplier No. 2234681KG

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Quantity 1 kg
UN Number 1490
Formula Weight 158.03g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Potassium Permanganate
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H302-H314-H410
EINECSNumber 231-760-3
RTECSNumber SD6475000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.