Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate Concentrate, 1/50M, For 1L standard solution, Solstice

Catalog No. 60046696 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Packaging:
Ampoule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-696 1 Pc. Ampoule
60-046-697 6 x 1 Ea. Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-696 Supplier Solstice Supplier No. 381301EA
Add to Cart
Add to Cart

for 1L standard solution, 1/50 M KMnO4 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Violet
Quantity 1 Pc.
UN Number UN3214
Formula Weight 158.03g/mol
Concentration or Composition (by Analyte or Components) 1/50 M KMnO4 (0.1N)
Packaging Ampoule
Appearance Clear liquid
Chemical Name or Material Potassium permanganate concentrate, For 1L standard solution, 1/50M KMnO4 (0.1N)
Hazard Statement P273-P501
DOTInformation 5.1/PGII
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.