Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, For Chlorine, Certified, 0.891g/L ±0.005, LabChem™

Catalog No. LC198602
Click to view available options
Quantity:
500 mL
1 L

Catalog No. LC198602

Supplier: LabChem, Inc. LC198602

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-64-7, 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Purple
CAS Max % 99.91, 0.09
Linear Formula KMnO4
Concentration 0.891g/L ±0.005
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 158.04
Grade Certified
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Manganese; Oxygen
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Permanganate, For Chlorine
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.