Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium permanganate, Powder, crystals or chunks, ≤150 μm particle size (DLS), 97%, Solstice

Catalog No. 60045472 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-472 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-472 Supplier Solstice Supplier No. 238511500G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Powder, crystals or chunks, ≤150 μm particle size (DLS), 97%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

CAS Max % 100.0000%
Physical Form Powder
Quantity 500 g
Linear Formula KMnO4
UN Number UN1490
Formula Weight 158.03
Percent Purity 97%
Packaging Glass Bottle
Density 2.700 g/cm3
Chemical Name or Material Potassium permanganate, Powder, crystals or chunks, ≤150 μm particle size (DLS), 97%
Hazard Statement P220-P273-P280-P305 + P351 + P338-P310-P501
Precautionary Statement H272-H302-H314-H410
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 5.1; Packing Group:3: II; Proper Shipping Name: Potassium permanganate
EINECSNumber 231-760-3
RTECSNumber SD6475000
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.