Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate Standard, 891 mg/L (1000 ppm as Cl2), Ricca Chemical
SDP

Catalog No. R63543001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R63543001C 1 L Amber Glass Bottle
R6354300500 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R63543001C Supplier Ricca Chemical Company Supplier No. R63543001C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 891mg/L
CAS Min % 98.0, 0.09
CAS Max % 100.0, 0.09
Physical Form Liquid
Color Purple
Boiling Point 100°C
Quantity 1 L
Chemical Name or Material Potassium Permanganate Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.