Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Persulfate, ACS Grade, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88704 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88704 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88704 Supplier Aqua Phoenix Scientific Supplier No. PP1060J
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS-grade potassium persulfate suitable for laboratory chemical applications.

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.31
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
IUPAC Name dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Powder
Quantity 100 g
Grade ACS
Chemical Name or Material Potassium Persulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.