Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 7.0

Catalog No. AAJ61261K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61261K2 1 L
AAJ61261AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61261K2 Supplier Thermo Scientific Chemicals Supplier No. J61261K2
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7778-77-0 | H2KO4P | 136.084 g/mol

Potassium phosphate is typically used as a component in the preparation of phosphate based buffers. It is convenient stock solution for making phosphate buffers. The pKa of 6.82 (25 C) for phosphate buffers make them a universal buffer for biological applications such as cell culture media, column eluent solutions and reagents used in molecular biology. Specific applications include buffers for protein purification, bioprocessing and diagnostic reagent production. Potassium salts are used where sodium content needs to be minimized or controlled.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate is typically used as a component in the preparation of phosphate based buffers. It is convenient stock solution for making phosphate buffers. The pKa of 6.82 (25 C) for phosphate buffers make them a universal buffer for biological applications such as cell culture media, column eluent solutions and reagents used in molecular biology. Specific applications include buffers for protein purification, bioprocessing and diagnostic reagent production. Potassium salts are used where sodium content needs to be minimized or controlled.

Solubility
Fully miscible in water. Insoluble in ethanol.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.084
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Concentration 0.2M buffer soln.
pH 7.0
Physical Form Liquid
Quantity 1 L
Recommended Storage Ambient Temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.