Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic, ACS Reagent (Potassium Dihydrogen Phosphate), Reagents

Catalog No. 01337799
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-799 500 g Glass Bottle
01-337-803 100 g Glass Bottle
01-337-802 10 kg Poly Pail
01-337-801 3 kg Poly Bottle
01-337-800 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-799 Supplier Reagents Holdings Llc Supplier No. C1289000500C2
Add to Cart
Add to Cart

Potassium Phosphate Monobasic, ACS Reagent (Potassium Dihydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Phosphate Monobasic, ACS Reagent (Potassium Dihydrogen Phosphate)
Boiling Point 400°C (loses water and becomes metaphosphate)
Melting Point 253°C
Color Colorless/White
CAS 7778-77-0
pH 4.1 to 4.5
Physical Form Solid
Quantity 500 g
Molecular Formula KH2PO4
Synonym Potassium Dihydrogen Phosphate
Molecular Weight (g/mol) 136.0855
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.338
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.