Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg

Catalog No. 5295681KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-956-81KG 1 kg
52-956-8250GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-956-81KG Supplier MilliporeSigma™ Supplier No. 5295681KG
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Potassium Phosphate, Monobasic
CAS 7778-77-0
Quantity 1 kg
Purity Grade Notes by titration
Assay Percent Range ≥99%
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Formula Weight 136.1
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.