Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Tribasic, FCC, 97%, Spectrum™ Chemical
SDP

Catalog No. 18605687 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
20 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-687 45 kg Fiber Drum
18-605-683 125 g Poly Bottle
18-605-684 500 g Poly Bottle
18-605-685 2.5 kg Poly Pail
18-605-686 12 kg Fiber Drum
18-605-688 20 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-687 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P137945KGBL
Add to Cart
Add to Cart
Request a Quote

P1379, 7778-53-2, K3PO4

Spectrum™ Chemical Potassium Phosphate Tribasic, FCC is used as a food additive for its properties as an emulsifier, foaming agent, and whipping agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.27
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
IUPAC Name tripotassium phosphate
SMILES [K+].[K+].[K+].[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 45 kg
Percent Purity 97%
Grade FCC
Loss on Ignition 0.05
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.