Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phthalate Monobasic, ≥99.5% (T), For HPLC, Solstice

Catalog No. 60047778 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-778 50 g Poly Bottle
60-047-777 250 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-778 Supplier Solstice Supplier No. 9614850G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for HPLC,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Boiling Point 378°C
Melting Point 295°C to 300°C
Color Colorless
pH 3.5 to 5
Physical Form Solid
Quantity 50 g
Linear Formula HOOCC6H4COOK
UN Number NONH for all modes of transport
Beilstein 3637128
Formula Weight 204.22g/mol
Percent Purity ≥99.5% (T)
Loss on Drying max. 0.5% (105°C)
Odor Odorless
Packaging Poly Bottle
Density 1.64g/cm3
Chemical Name or Material Potassium phthalate monobasic
Show More Show Less
RTECSNumber 212-889-4
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.