Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium silicate, anhydrous

Catalog No. p-4407762
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4449322 100 g
AA4449336 500 g
AA44493A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4449322 Supplier Thermo Scientific Chemicals Supplier No. 04449322
Only null left
Add to Cart

CAS: 1312-76-1 | K2O3Si | 154.28 g/mol

Potassium silicate is used in binders, carbon electrodes, lead pencils, protective coatings, insoluble pigments, phosphors on television tubes and detergent additives. It acts as a catalyst to make glass, ceramics, welding rods, adhesives, cements, gels, paints, pulp and paper production. Further, it is used in horticulture as a soluble source of potassium and silicon. It acts as a corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-76-1
Molecular Formula K2O3Si
Molecular Weight (g/mol) 154.28
MDL Number MFCD00145152
InChI Key NNHHDJVEYQHLHG-UHFFFAOYSA-N
IUPAC Name dipotassium oxosilanebis(olate)
SMILES [K+].[K+].[O-][Si]([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water.
Odor Odorless
Chemical Name or Material Potassium silicate
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 215-199-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.