Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate, Tetrahydrate, Purified, 95%, Spectrum™ Chemical
SDP

Catalog No. 18605708 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-708 2.5 kg Poly Pail
18-605-707 500 g Poly Bottle
18-605-709 12 kg Poly Pail
18-605-710 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-708 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14012.5KG
Add to Cart
Add to Cart

P1401, 6381-59-5, C4H4KNaO6·4H2O

Spectrum™ Chemical Potassium Sodium Tartrate, Tetrahydrate, Purified was one of the first materials discovered to exhibit piezoelectricity. It is a double salt and is deliquescent so the material deteriorates if stored in damp conditions The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Chemical Identifiers

CAS 6381-59-5
Molecular Formula C4H12KNaO10
Molecular Weight (g/mol) 282.22
InChI Key VZOPRCCTKLAGPN-ZFJVMAEJSA-L
IUPAC Name potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
SMILES O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 95%
Grade Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.