Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Tellurite MP Biomedicals

Catalog No. icn10042725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10042725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10042725 Supplier MP Biomedicals, Inc Supplier No. 0210042725
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7790-58-1
Molecular Formula K2O3Te
Molecular Weight (g/mol) 253.794
InChI Key BFPJYWDBBLZXOM-UHFFFAOYSA-L
Synonym potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e
PubChem CID 65186
ChEBI CHEBI:75248
IUPAC Name dipotassium;tellurite
SMILES [O-][Te](=O)[O-].[K+].[K+]

Specifications

Quantity 25 g
Formula Weight 253.8
Chemical Name or Material Potassium Tellurite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.