Learn More
Selleck Chemical LLC Abemaciclib mesylate
SDP
Supplier:  Selleck Chemical LLC S7158100MG
Encompass_Preferred
Abemaciclib mesylate is a potent and selective inhibitor of CDK4 and CDK6 with IC50 of 2 nM and 10 nM in cell-free assays respectively Phase 3
Catalog No. 50-000-04267
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.