Learn More
Potassium thiocyanate, 98%
Description
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.
Solubility
Soluble in water, acetone and alcohol.
Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.
Specifications
Specifications
| Melting Point | 172°C to 175°C |
| Density | 1.89 |
| pH | 5.3 to 8.5 |
| Boiling Point | 500°C (decomposition) |
| Odor | Odorless |
| Quantity | 2500 g |
| Beilstein | 3594799 |
| Sensitivity | Hygroscopic; Light sensitive |
| Merck Index | 14,7691 |
| Solubility Information | Soluble in water,acetone and alcohol. |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| EINECSNumber | 206-370-1 |
| RTECSNumber | XL1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.