Learn More
Specifications
Specifications
| Color | Colorless or White |
| CAS Min % | 98.0 |
| pH | 5.3 to 8.7 |
| CAS Max % | 100.0 |
| Physical Form | Crystals |
| Quantity | 10 kg |
| Synonym | potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt |
| SMILES | C(#N)[S-].[K+] |
| IUPAC Name | potassium;thiocyanate |
| Molecular Weight (g/mol) | 97.176 |
| Show More |
Safety and Handling
| ShelfLife | 60 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
