Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium trifluoromethanesulfonate, 98%

Catalog No. p-7049731
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2036109 10 g
AA2036118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2036109 Supplier Thermo Scientific Chemicals Supplier No. 02036109
Only null left
Add to Cart

CAS: 2926-27-4 | CF3KO3S | 188.162 g/mol

Potassium trifluoromethanesulfonate is used as a reagent in the synthesis of guanine-quadruplex hybrid materials. It acts as a supporting electrolyte in the electrochemical study of evidence for gold anion in ethylenediamine. It is involved in the preparation of N-fluoro-2,4,6-trimethylpyridinium triflate by reacting with 2,4,6-trimethyl-pyridine. Further, it is used in the preparation of imidazolium salt, 3-methyl-1-(3R,3aR,6S,6aR)-[6-(benzyloxy)-hexahydrofuro[3,2-b]furan-3-yl]imidazolium trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2926-27-4
Molecular Formula CF3KO3S
Molecular Weight (g/mol) 188.162
MDL Number MFCD00042370
InChI Key GLGXXYFYZWQGEL-UHFFFAOYSA-M
Synonym potassium trifluoromethanesulfonate, potassium triflate, trifluoromethanesulfonic acid potassium salt, potassium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, potassium salt, potassium ion triflate, cf3so3k, acmc-209h7u, potassium trifluoromethanesulfonate salt, potassium tris fluoranyl methanesulfonate
PubChem CID 23664779
IUPAC Name potassium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[K+]

Specifications

Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 188.17
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Potassium trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.