Learn More
Sigma Organic Chemistry Potassium hexacyanoferrate(II) trihydrate | 100G | 14459-95-1 | MFCD00167023 | >=99.95% trace metals basis
Supplier: Sigma Organic Chemistry 455989100G

Potassium hexacyanoferrate(II) trihydrate, 100G
About This Item:
Storage: room temp
EINECS Number: 237-722-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.