Learn More
Bachem (D-Ala1)-Peptide T amide
SDP
Supplier:  Bachem 4015906.0005
Encompass_Preferred
(D-Ala1)-Peptide T amide, H-D-Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr-NH2, 106362-34-9, C35H56N10O15, Mr 856.89, 5mg. DAPTA. Potent analog of peptide T. DAPTA (aSTTTNYT-amide) showed a strong anti-HIV 1 activity in monocytes/macrophages, the peptide inhibits the entry of the virus.
Catalog No. 50-194-6949