Learn More
Bachem Endothelin-3 (human, mouse, rabbit, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4095915.0500
Encompass_Preferred
Endothelin-3 (human, mouse, rabbit, rat) Trifluoroacetate, H-Cys-Thr-Cys-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys-Val-Tyr-Tyr-Cys-His-Leu-Asp-Ile-Ile-Trp-OH, 117399-93-6, C121H168N26O33S4, Mr 2643.08, 0.5mg. ET-3 (human, mouse, rabbit, rat), Endothelin-3, human. Potent constrictor of rat blood vessels in vitro and an effective pressor agent in vivo. Endothelin-3 (ET-3) has been found in high concentrations in the brain and may regulate important functions in neurons and astrocytes, such as proliferation and development. ET-3 displays selectivity for the ETC receptor.
Catalog No. 50-194-6780