Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU3F4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734729 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4729 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4729 Supplier Proteintech Group Inc Supplier No. 251141AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.

Specifications

Antigen POU3F4
Applications Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene POU3F4
Gene Accession No. P49335, P62515
Gene Alias BRAIN 4, Brn 4, BRN4, DFN3, DFNX2, Oct 9, Octamer binding protein 9, OTF 9, OTF9, POU class 3 homeobox 4, POU3F4
Gene Symbols Pou3f4
Host Species Rabbit
Immunogen POU3F4 Fusion Protein Ag18925
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18994, 5456
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.