Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU4F1 Recombinant Rabbit Monoclonal Antibody (9A5O9)

Catalog No. MA557983
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA557983 20 μL
MA557984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA557983 Supplier Invitrogen™ Supplier No. MA557983
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MMSMNSKQPH FAMHPTLPEH KYPSLHSSSE AIRRACLPTP PLQSNLFASL DETLLARAEA LAAVDIAVSQ GKSHPFKPDA TYHTMNSVPC TSTSTVPLAH.

BRN3A is a class IV POU domain-containing transcription factor highly expressed in the developing sensory nervous system and in cells of the B- and T-lymphocytic lineages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU4F1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9A5O9
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Pou4f1
Gene Accession No. Q01851
Gene Alias Brain-3A; brain-specific homeobox/POU domain protein 3A; Brn3; Brn-3; brn3.0; Brn-3.0; Brn3a; brn-3A; brn3b; E130119J07Rik; etID22526.1; FLJ13449; homeobox/POU domain protein RDC-1; LOW QUALITY PROTEIN: POU domain, class 4, transcription factor 1; Oct-T1; POU class 4 homeobox 1; POU domain, class 4, transcription factor 1; POU4F1; pou4f1 protein; RDC1; RDC-1
Gene Symbols Pou4f1
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5457
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.