Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Povidone-Iodine, USP, 9-12%, Spectrum™ Chemical
SDP

Catalog No. 18605760 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-760 12 kg Fiber Drum
18-605-757 100 g Amber Glass Bottle
18-605-758 500 g Amber Glass Bottle
18-605-759 2.5 kg Poly Pail
18-605-761 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-760 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO33012KG
Add to Cart
Add to Cart

PO330, 25655-41-8, (C8H9NO)n·xI

Spectrum™ Chemical Povidone-Iodine, USP is used as a broad spectrum antiseptic for topical application in the treatment and prevention of infection in wounds. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25655-41-8
Molecular Formula (C6H9NO)nI2
Molecular Weight (g/mol) 364.95
MDL Number MFCD00084483
InChI Key CPKVUHPKYQGHMW-UHFFFAOYSA-N
IUPAC Name 1-ethenylpyrrolidin-2-one; diiodine
SMILES II.*-CC(-*)N1CCCC1=O

Specifications

CAS Max % 1
Ignition Residue 0.00025
Loss on Drying 0.08
Packaging Fiber Drum
Quantity 12 kg
Percent Purity 9.0 to 12%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.