Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Povidone, K-90, USP, Spectrum™ Chemical
SDP

Catalog No. 18605752 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-752 100 g Poly Bottle
18-605-754 2.5 kg Poly Bottle
18-605-755 12 kg Poly Bottle
18-605-756 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-752 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1416100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1416, 9003-39-8, (C6H9NO)n

Spectrum™ Chemical Povidone, K-90, USP is used as a binder in pharmaceutical tablets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

CAS Max % 1
pH 4.0 to 7.0
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 100 g
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Povidone K-90
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.