Learn More
BURKLE INC Telescopic rod, infinitely adj. 27.56–70.87 in. (70–180 cm), 4 rods
Supplier:  BURKLE INC 53550180
Encompass_Preferred
Different tools can be attached to the telescopic rod (snap-in joint). The individual rods have been provided with an end stop. This means that one rod cannot accidentally slip out of another rod.
The telescopic rods cannot be rotated radially, which means even better handling!
Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm (110.24 in.). Fits into every car trunk!
Catalog No. 50-000-25781
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.