Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Powdered Latex Finger Cots

Catalog No. 19055210
Change view
Click to view available options
Size:
Medium
X-Large
Packaging Type:
Bulk
Standard
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19-055210 Medium Standard
19-069952 Medium Bulk
19-069937 X-Large Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-055210 Supplier Honeywell™ North™ Supplier No. 321LWR/M

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Thin gauge ensures an extremely high level of tactile sensitivity, with a tapered design for improved fit and comfort.

  • Thin gauge (3 mil) ensures extremely high levels of tactile sensitivity
  • Tapered design improves fit and comfort
  • Compliant with USDA and FDA regulations, 21 CFR, for use in food processing
  • Available in standard or bulk packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA Standard - 29 CFR 1915.157
Color White
Disposable Single-use
Exterior Finish No Texture
Hand Ambidextrous
Material Latex
Powdered Powdered
Product Type Finger Cot
Size Medium
Thickness (English) 3 mil
For Use With (Application) Electrical Work, Food Processing, Photo Processing, and Battery/Watch Manufacturing
Glove Cuff Style Rolled
Packaging Type Standard
Length (English) 2.8 in.
Length (Metric) 7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.