Learn More
Vernier™ Power Amplifier
SDPSCI_ED
Catalog No. S15958ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Educational use only
Catalog No. For Use With (Application)
S15958ND Educational use only
1 options
Catalog No. S15958ND Supplier Vernier™ Supplier No. PAMP
Add to Cart
Add to Cart
Includes
Vernier Power Amplifier, (1) Power supply, (1) 3.5 mm audio cable for LabQuest control, (1) BTA cable for LabPro input and current sensing
Requires
The Power Amplifier's current sensor requires a compatible interface and software for data logging.
Use the Power Amplifier as a power supply for DC and ACcircuit investigations or to drive devices such as speakers, lamps, and small DC motors.
Technical Details of Power Amplifier Operation Input connection: 3.5 mm audio cable (from LabQuest or computer) or BTA cable (from LabPro CH4) Input Frequency range: DC to 15 kHz. Overall gain: 10X (LabQuest/computer), 2X (LabPro) Input impedance: 10 kΩ. (LabQuest/computer), 50 kΩ (LabPro) Maximum input ±12 V without damage. Pulse Width Modulation input used for producing DC offset or pure DC output Current Sensor Connect to Vernier interface to monitor load current There is a 0.1 Ω resistor between the black terminal and ground. Current measurements are based on the voltage drop across this resistor. Important Note: The Power Amplifier creates no signal on its own. It depends on a signal source, such as the LabQuest 2 or original LabQuest’s Power Amplifier App, our free Power Amplifier Frequency Generator software for PC/Mac, or Logger Pro 3.8 or newer (LabPro only) A LabQuest Mini or LabQuest Stream does not provide a signal source. Use the Power Amplifier Function Generator software as the signal source.
Warranty and Services
5-year limited warranty
Specifications
Description Depends on a signal source
Requirements Compatible interface and software data for logging
For Use With (Application) Educational use only
For Use With (Equipment) LabQuest 2 or Original LabQuest's Power Amplifier App
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov