Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Power Blotter 1-Step™ Transfer Buffer (5X)

Catalog No. p-7180366
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PB7100 250 mL
PB7300 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PB7100 Supplier Invitrogen™ Supplier No. PB7100
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen Power Blotter 1-Step Transfer Buffer is designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Power Blotter System.

Invitrogen Power Blotter 1-Step Transfer Buffer is designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Power Blotter System. Transfer is compatible with commonly used detection methods such as staining, chemiluminescence, or fluorescence.

Features of Power Blotter 1-Step Transfer Buffer:
Fast—high ionic strength formulation allows for 5- to 12-minute protein transfer when used with compatible fast semi-dry blotting systems
Optimized—designed for seamless and reliable performance with the Power Blotter System
Compatible—effective with other semi-dry transfer devices equipped with a suitable high-current power supply
Economical—no need for special or costly consumables that are required by other fast semi-dry transfer devices

When used with the Power Blotter System, this buffer provides effective gel-to-membrane transfer of proteins in 5 to 12 minutes with efficiency that is similar to conventional western blotting techniques. Fast blotting methods require a high-current power supply, such as the Power Blotter System, and an optimized high ionic strength transfer buffer, such as Power Blotter 1-Step Transfer Buffer.

By increasing the current, excellent transfer efficiency can be achieved in much shorter time compared to conventional methods. Amperage is held at a constant rate based on the surface area of the transfer stack(s) (∼22–23 mA/cm2) and voltage is limited to 25 V.

Power Blotter 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices, when they are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.

Related products
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular size (Cat. No. PB3310)
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, mini (PB3210)
Power Blotter Pre-cut Membranes and Filters, PVDF, regular size (PB5310)
Power Blotter Pre-cut Membranes and Filters, PVDF, mini (PB4210)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Semi-Dry
Product Line 1-Step
Type Transfer Buffer
Quantity 250 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.